(3S)-(-)-3-(Dimethylamino)pyrrolidine ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
(S)-3-(Dimethylamino)pyrrolidine, (S)-(-)-3-(Dimethylamino)pyrrolidine
D2193-1G
D2193-5G
TCIAD2193-1GEA
1375
NOK
TCIAD2193-1G
TCIAD2193-5G
(3S)-(-)-3-(Dimethylamino)pyrrolidine ≥98.0% (by GC)
(3S)-(-)-3-(Dimethylamino)pyrrolidine
Formel:
C₆H₁₄N₂ MW: 114,19 g/mol Kokepunkt: 164°C Flash punkt: 51°C |
MDL nummer:
MFCD00191348 CAS-nummer: 132883-44-4 FN: 2734 ADR: 8,II |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -13.0 to -16.0 deg (C=10, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...