(-)-1,4-Di-O-benzyl-L-threitol ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
(-)-(2S,3S)-1,4-Bis(benzyloxy)-2,3-butanediol
D2240-1G
D2240-5G
TCIAD2240-1GEA
1425
NOK
TCIAD2240-1G
TCIAD2240-5G
(-)-1,4-Di-O-benzyl-L-threitol ≥98.0% (by GC)
(-)-1,4-Di-O-benzyl-L-threitol
Formel:
C₁₈H₂₂O₄ MW: 302,37 g/mol |
MDL nummer:
MFCD00077723 CAS-nummer: 17401-06-8 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 56.0 to 59.0 deg-C |
Specific rotation [a]20/D | -6.0 to -6.5 deg (C=5, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...