3,4-Dimethylhippuric acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
3,4-Dimethylhippuric acid, 2-(3,4-Dimethylbenzamido)acetic acid, N-(3,4-Dimethylbenzoyl)glycine
D2241-1G
D2241-5G
D2241-200MG
TCIAD2241-1GEA
13325
NOK
TCIAD2241-1G
TCIAD2241-5G
TCIAD2241-200MG
3,4-Dimethylhippuric acid ≥98.0% (by HPLC, titration analysis)
3,4-Dimethylhippuric acid
Formel:
C₁₁H₁₃NO₃ MW: 207,23 g/mol |
MDL nummer:
MFCD00191414 CAS-nummer: 23082-12-4 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 159.0 to 162.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...