Diphenyliodonium perchlorate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

D2243-1G D2243-5G
TCIAD2243-1GEA 1675 NOK
TCIAD2243-1G TCIAD2243-5G
Diphenyliodonium perchlorate ≥98.0% (by titrimetric analysis)
Diphenyliodonium perchlorate
Formel: C₁₂H₁₀ClIO₄
MW: 380,56 g/mol
MDL nummer: MFCD00191355
CAS-nummer: 75007-13-5
FN: 1479
ADR: 5.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Melting point 175.0 to 177.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR