(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥98.0% (by HPLC) HPLC labeling reagent for e.e. determination
Leverandør: TCI
|
Warning
|
Synonymer:
1-Fluoro-2,4-dinitrophenyl-5-L-alanine amide, N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, Propanamide, 2-[(5-fluoro-2,4-dinitrophenyl)amino]-, (2S)-
D2259-100MG
D2259-1G
TCIAD2259-100MGEA
1000
NOK
TCIAD2259-100MG
TCIAD2259-1G
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥98.0% (by HPLC) HPLC labeling reagent for e.e. determination
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide
Formel:
C₉H₉FN₄O₅ MW: 272,192 g/mol |
MDL nummer:
MFCD00042049 CAS-nummer: 95713-52-3 |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 99.0 ee% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...