5-Amino-6-chloro-2,3-dicyanopyrazine ≥98.0% (by GC, total nitrogen)

Leverandør: TCI
Danger

Synonymer: 5-Amino-6-chloro-2,3-pyrazinedicarbonitrile, 5-Amino-6-chloropyrazine-2,3-dicarbonitrile

D2269-10G D2269-1G
TCIAD2269-10GEA 3600 NOK
TCIAD2269-10G TCIAD2269-1G
5-Amino-6-chloro-2,3-dicyanopyrazine ≥98.0% (by GC, total nitrogen)
5-Amino-6-chloro-2,3-dicyanopyrazine
Formel: C₆H₂ClN₅
MW: 179,57 g/mol
Smeltepunkt: 208°C
MDL nummer: MFCD00191417
CAS-nummer: 56413-96-8
FN: 2811
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 206.0 to 210.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR