2,4-Dimethoxypyrimidine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
D2327-25G D2327-5G
TCIAD2327-25GEA 1500 NOK
TCIAD2327-25G TCIAD2327-5G
2,4-Dimethoxypyrimidine ≥98.0% (by GC, titration analysis)
2,4-Dimethoxypyrimidine
Formel: C₆H₈N₂O₂
MW: 140,14 g/mol
Kokepunkt: 205°C
Flash punkt: 92°C
MDL nummer: MFCD00056028
CAS-nummer: 3551-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e4e60babb901d30fb7208eb3b27f1625797e351&vid=6e4e60babb901d30fb7208eb3b27f1625797e351&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %