N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine ≥98.0% (by total nitrogen basis)

Leverandør: TCI

Synonymer: 4,4'-Bis[N-phenyl-N-(m-tolyl)amino]biphenyl, N,N'-Diphenyl-N,N'-di(m-tolyl)benzidine, TPD, N,N'-Bis(3-methylphenyl)-N,N'-diphenylbenzidine

D2448-1G D2448-5G
TCIAD2448-1GEA 300 NOK
TCIAD2448-1G TCIAD2448-5G
N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine ≥98.0% (by total nitrogen basis)
N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine
Formel: C₃₈H₃₂N₂
MW: 516,6741 g/mol
MDL nummer: MFCD00144965
CAS-nummer: 65181-78-4
EINECS: 413-810-8
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR