Dimethyl-2,5-dioxahexanedioate ≥98.0% (by GC)

Leverandør: TCI

D2455-25G D2455-5G
TCIAD2455-25GEA 2075 NOK
TCIAD2455-25G TCIAD2455-5G
Dimethyl-2,5-dioxahexanedioate ≥98.0% (by GC)
Dimethyl-2,5-dioxahexanedioate
Formel: C₆H₁₀O₆
Kokepunkt: 220°C
MDL nummer: MFCD00085447
CAS-nummer: 88754-66-9

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR