Diethylene glycol bis(p-toluenesulphonate) ≥96.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Oxybis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), Diethylene Glycol Ditosylate, Bis(2-tosyloxyethyl) Ether, Bis[2-(p-toluenesulphonyloxy)ethyl] Ether
D2457-25G
D2457-5G
TCIAD2457-25GEA
1200
NOK
TCIAD2457-25G
TCIAD2457-5G
Diethylene glycol bis(p-toluenesulphonate) ≥96.0% (by HPLC, titration analysis)
Diethylene glycol bis(p-toluenesulphonate)
Formel:
C₁₈H₂₂O₇S₂ MW: 414,49 g/mol |
MDL nummer:
MFCD00026001 CAS-nummer: 7460-82-4 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Ester Value) | min. 96.0 % |
Melting point | 85.0 to 89.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...