Diphenyliodonium-2-carboxylate monohydrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: (2-Carboxyphenyl)phenyliodonium Hydroxide Inner Salt

D2503-25G D2503-5G
TCIAD2503-25GEA 8975 NOK
TCIAD2503-25G TCIAD2503-5G
Diphenyliodonium-2-carboxylate monohydrate ≥98.0% (by HPLC, titration analysis)
Diphenyliodonium-2-carboxylate monohydrate
Formel: C₁₃H₉IO₂
MW: 324,12 g/mol
MDL nummer: MFCD00150328
CAS-nummer: 96195-89-0
EINECS: 216-070-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e07714767e0955055dd6349c4d81d6b2b6e978&vid=02e07714767e0955055dd6349c4d81d6b2b6e978&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 4.0 to 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR