(2,3-Dichlorophenyl)acetonitrile ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
2,3-Dichlorophenylacetonitrile, 2-(2,3-Dichlorophenyl)acetonitrile, 2,3-Dichlorobenzyl cyanide
D2945-25G
D2945-5G
TCIAD2945-25GEA
1800
NOK
TCIAD2945-25G
TCIAD2945-5G
(2,3-Dichlorophenyl)acetonitrile ≥98.0% (by GC)
(2,3-Dichlorophenyl)acetonitrile
Formel:
C₈H₅Cl₂N MW: 186,04 g/mol Kokepunkt: 115°C |
MDL nummer:
MFCD01861392 CAS-nummer: 3218-45-9 FN: 3439 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...