Di-n-octyl sulphoxide ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: Di-n-octylsulphoxide, n-Octyl Sulphoxide

D2965-5G D2965-1G
TCIAD2965-5GEA 1925 NOK
TCIAD2965-5G TCIAD2965-1G
Di-n-octyl sulphoxide ≥98.0% (by HPLC)
Di-n-octyl sulphoxide
Formel: C₁₆H₃₄OS
MW: 274,51 g/mol
MDL nummer: MFCD00039514
CAS-nummer: 1986-89-6
EINECS: 217-861-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR