1,3,2-Dioxathiolane-2-oxide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Ethylene Sulphite, Glycol sulphite

D2977-25G D2977-5G
TCIAD2977-25GEA 500 NOK
TCIAD2977-25G TCIAD2977-5G
1,3,2-Dioxathiolane-2-oxide ≥98.0% (by GC)
1,3,2-Dioxathiolane-2-oxide
Formel: C₂H₄O₃S
MW: 108,11 g/mol
Kokepunkt: 172°C
Flash punkt: 114°C
MDL nummer: MFCD00005354
CAS-nummer: 3741-38-6
EINECS: 223-131-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR