2,2'-Dinaphthylamine ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,2'-Iminodinaphthalene

D2987-1G D2987-5G
TCIAD2987-1GEA 700 NOK
TCIAD2987-1G TCIAD2987-5G
2,2'-Dinaphthylamine ≥98.0% (by GC)
2,2'-Dinaphthylamine
Formel: C₂₀H₁₅N
MW: 269.35 g/mol
Kokepunkt: 471°C
MDL nummer: MFCD03093264
CAS-nummer: 532-18-3
EINECS: 208-529-0
Merck Indeks: 14,03267

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR