3,6-Diphenyl-1,2,4,5-tetrazine ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Warning

D3175-1G D3175-5G
TCIAD3175-1GEA 755 NOK
TCIAD3175-1G TCIAD3175-5G
3,6-Diphenyl-1,2,4,5-tetrazine ≥98.0% (by total nitrogen basis)
3,6-Diphenyl-1,2,4,5-tetrazine
Formel: C₁₄H₁₀N₄
Smeltepunkt: 196°C
MDL nummer: MFCD00023646
CAS-nummer: 6830-78-0

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR