N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
N-(4-Methyl-2-pentyl)-N'-phenyl-1,4-phenylenediamine, 4-[(4-Methyl-2-pentyl)amino]diphenylamine, N1-(4-Methylpentan-2-yl)-N4-phenylbenzene-1,4-diamine
D3331-500G
D3331-25G
TCIAD3331-500GEA
1775
NOK
TCIAD3331-500G
TCIAD3331-25G
N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine ≥98.0% (by GC)
N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine
Formel:
C₁₈H₂₄N₂ MW: 268,40 g/mol Kokepunkt: 165°C |
MDL nummer:
MFCD00072248 CAS-nummer: 793-24-8 EINECS: 212-344-0 FN: 3077 ADR: 9,III |
Spesifikasjon Testresultater
Appearance | Red to Dark blue to Black pellet |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...