Diethylene glycol monobenzyl ether ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
2-[2-(Benzyloxy)ethoxy]ethanol, 2-(2-(Benzyloxy)ethoxy)ethanol
D3408-25G
D3408-500G
TCIAD3408-25GEA
220
NOK
TCIAD3408-25G
TCIAD3408-500G
Diethylene glycol monobenzyl ether ≥98.0% (by GC)
Diethylene glycol monobenzyl ether
Formel:
C₁₁H₁₆O₃ MW: 196,25 g/mol Flash punkt: 158°C |
MDL nummer:
MFCD00029649 CAS-nummer: 2050-25-1 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...