4-(Dimethylamino)benzyl alcohol ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Hydroxymethyl-N,N-dimethylaniline
D3482-25G
D3482-5G
TCIAD3482-25GEA
8600
NOK
TCIAD3482-25G
TCIAD3482-5G
4-(Dimethylamino)benzyl alcohol ≥98.0% (by titrimetric analysis)
4-(Dimethylamino)benzyl alcohol
Formel:
C₉H₁₃NO MW: 151,21 g/mol Kokepunkt: 123°C Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD03840449 CAS-nummer: 1703-46-4 |
Spesifikasjon Testresultater
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...