4-(Dimethylamino)butyraldehyde dimethyl acetal ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4,4-Dimethoxy-N,N-dimethylbutylamine

D3535-25G D3535-5G
TCIAD3535-25GEA 2375 NOK
TCIAD3535-25G TCIAD3535-5G
4-(Dimethylamino)butyraldehyde dimethyl acetal ≥98.0% (by GC)
4-(Dimethylamino)butyraldehyde dimethyl acetal
Formel: C₈H₁₉NO₂
MW: 161,25 g/mol
Kokepunkt: 54°C
MDL nummer: MFCD06411126
CAS-nummer: 19718-92-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR