DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon milt)

Leverandør: TCI

Synonymer: Thymonucleic acid sodium salt, DNA sodium salt

D3545-25G D3545-5G
TCIAD3545-25GEA 1825 NOK
TCIAD3545-25G TCIAD3545-5G
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon milt)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
MDL nummer: MFCD00130921
CAS-nummer: 9007-49-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Total Nitrogen 13.0 to 15.0 %
abs-ratio (a/b) min. 1.30 (E260/E280, in pH7.0 Phosphate buffer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR