3,3-Dimethyl-2-oxobutyric acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Danger
Synonymer: 3,3-Dimethyl-2-oxobutanoic acid, Trimethylpyruvic Acid
D3609-25G D3609-5G
TCIAD3609-25GEA 1300 NOK
TCIAD3609-25G TCIAD3609-5G
3,3-Dimethyl-2-oxobutyric acid ≥98.0% (by titrimetric analysis)
3,3-Dimethyl-2-oxobutyric acid
Formel: C₆H₁₀O₃
MW: 130,14 g/mol
Kokepunkt: 80°C
MDL nummer: MFCD00154352
CAS-nummer: 815-17-8
EINECS: 212-418-2
FN: 3265
ADR: 8,III
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %