Dithieno[3,2-b:2',3'-d]thiophene ≥98.0% (by GC)
Leverandør: TCI
D3641-1G
D3641-200MG
TCIAD3641-1GEA
3550
NOK
TCIAD3641-1G
TCIAD3641-200MG
Dithieno[3,2-b:2',3'-d]thiophene ≥98.0% (by GC)
Dithieno[3,2-b:2',3'-d]thiophene
Formel:
C₈H₄S₃ MW: 196,30 g/mol Kokepunkt: 138°C |
MDL nummer:
MFCD06656577 CAS-nummer: 3593-75-7 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...