3,3'-Dibromo-2,2'-bithiophene ≥98.0% (by GC)

Leverandør: TCI

D3798-1G D3798-25G D3798-5G
TCIAD3798-1GEA 500 NOK
TCIAD3798-1G TCIAD3798-25G TCIAD3798-5G
3,3'-Dibromo-2,2'-bithiophene ≥98.0% (by GC)
3,3'-Dibromo-2,2'-bithiophene
Formel: C8H4Br2S2
MW: 324,0554 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00114806
CAS-nummer: 51751-44-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR