1,1-Dibrom-3,3,3-trifluoraceton ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one

D4004-25G D4004-5G
TCIAD4004-25GEA 1300 NOK
TCIAD4004-25G TCIAD4004-5G
1,1-Dibrom-3,3,3-trifluoraceton ≥98.0% (by GC)
1,1-Dibrom-3,3,3-trifluoraceton
Formel: Br₂HCC(O)CF₃
MW: 269,84 g/mol
Kokepunkt: 113°C
MDL nummer: MFCD00041362
CAS-nummer: 431-67-4
FN: 3265
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR