2,5-Dibromo-4-picoline ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,5-Dibromo-4-methylpyridine

D4058-1G D4058-5G
TCIAD4058-1GEA 285 NOK
TCIAD4058-1G TCIAD4058-5G
2,5-Dibromo-4-picoline ≥98.0% (by GC)
2,5-Dibromo-4-picoline
Formel: C₆H₅Br₂N
MW: 250,92 g/mol
MDL nummer: MFCD00234955
CAS-nummer: 3430-26-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR