Donepezil hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 2-(1-Benzyl-4-piperidylmethyl)-5,6-dimethoxy-1-indanone Hydrochloride, 2-((1-Benzylpiperidin-4-yl)methyl)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one hydrochloride

D4099-5G D4099-1G
TCIAD4099-5GEA 2150 NOK
TCIAD4099-5G TCIAD4099-1G
Donepezil hydrochloride ≥98.0% (by HPLC, titration analysis)
Donepezil hydrochloride
Formel: C₂₄H₂₉NO₃
MW: 415,96 g/mol
MDL nummer: MFCD00881312
CAS-nummer: 120011-70-3
FN: 2811
ADR: 6.1,II
Merck Indeks: 14,03419
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6264be500dce95f812776949409ed4993c3540f1&vid=6264be500dce95f812776949409ed4993c3540f1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR