3-(Dimethylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
Synonymer:
3-(N,N-Dimethylaminocarbonyl)phenylboronic acid, 3-(Dimethylcarbamoyl)benzeneboronic acid, (3-(Dimethylcarbamoyl)phenyl)boronic acid
D4194-1G
D4194-5G
TCIAD4194-1GEA
970
NOK
TCIAD4194-1G
TCIAD4194-5G
3-(Dimethylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Dimethylcarbamoyl)phenylboronic acid
Formel:
C₉H₁₂BNO₃ MW: 193,01 g/mol |
MDL nummer:
MFCD03412066 CAS-nummer: 373384-14-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...