2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 2-(3,5-Dichloro-4-hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dichloro-4-hydroxyphenylboronic Acid Pinacol Ester

D4201-1G D4201-5G
TCIAD4201-1GEA 1875 NOK
TCIAD4201-1G TCIAD4201-5G
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97.0% (by GC, titration analysis)
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formel: C₁₂H₁₅BCl₂O₃
MW: 288,96 g/mol
Kokepunkt: 103°C
MDL nummer: MFCD20526385
CAS-nummer: 1003298-87-0

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR