3,6-Diklor-4-metylpyridazin ≥98.0% (by GC)

Leverandør: TCI
Warning

D4206-1G D4206-5G
TCIAD4206-1GEA 395 NOK
TCIAD4206-1G TCIAD4206-5G
3,6-Diklor-4-metylpyridazin ≥98.0% (by GC)
3,6-Diklor-4-metylpyridazin
Formel: C₅H₄Cl₂N₂
MW: 163,01 g/mol
Kokepunkt: 151°C
MDL nummer: MFCD00006465
CAS-nummer: 19064-64-3
EINECS: 242-794-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR