(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: (4aS,7aS)-Octahydro-1H-pyrrolo[3,4-b]pyridine

D4210-1G D4210-5G
TCIAD4210-1GEA 660 NOK
TCIAD4210-1G TCIAD4210-5G
(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane ≥98.0% (by GC)
(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane
Formel: C₇H₁₄N₂
MW: 126,20 g/mol
Kokepunkt: 59°C
Flash punkt: 91°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD08458306
CAS-nummer: 151213-40-0

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -5.0 to -8.0 deg (C=0.5, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR