[(2,6-Dimethylphenyl)amino](oxo)acetic Acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 2,6-Dimethylanilino(oxo)acetic acid, 2-((2,6-Dimethylphenyl)amino)-2-oxoacetic acid, N-(2,6-Dimethylphenyl)oxamic acid, DMPAO, (2,6-Dimethylanilino)(oxo)acetic acid, [(2,6-Dimethylphenyl)Carbamoyl]formic acid

D4265-1G D4265-5G
TCIAD4265-1GEA 1475 NOK
TCIAD4265-1G TCIAD4265-5G
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid ≥98.0% (by HPLC, titration analysis)
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid
Formel: C₁₀H₁₁NO₃
MW: 193,20 g/mol
MDL nummer: MFCD10018453
CAS-nummer: 2903-48-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR