2,4-Dibromobenzoic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

D4278-1G D4278-5G
TCIAD4278-1GEA 650 NOK
TCIAD4278-1G TCIAD4278-5G
2,4-Dibromobenzoic acid ≥98.0% (by GC, titration analysis)
2,4-Dibromobenzoic acid
Formel: C₇H₄Br₂O₂
MW: 279,92 g/mol
MDL nummer: MFCD00234253
CAS-nummer: 611-00-7
EINECS: 210-245-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 171.0 to 175.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR