2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Synonymer:
2,6-Dichloro-7-deazapurine
D4284-1G
D4284-5G
TCIAD4284-1GEA
730
NOK
TCIAD4284-1G
TCIAD4284-5G
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine ≥98.0% (by HPLC, total nitrogen)
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
Formel:
C₆H₃Cl₂N₃ MW: 188,01 g/mol |
MDL nummer:
MFCD08059278 CAS-nummer: 90213-66-4 |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 249.0 to 253.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...