N,N-Dimethyl-1,4-cyclohexanediamine (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 1-Amino-4-(dimethylamino)cyclohexane

D4288-1G D4288-5G
TCIAD4288-1GEA 1200 NOK
TCIAD4288-1G TCIAD4288-5G
N,N-Dimethyl-1,4-cyclohexanediamine (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-1,4-cyclohexanediamine (cis and trans mixture)
Formel: C₈H₁₈N₂
MW: 142,25 g/mol
Kokepunkt: 205°C
MDL nummer: MFCD06658434
CAS-nummer: 42389-50-4
FN: 2735
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR