5,6-Dichlorobenzimidazole Hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Warning
D4295-1G D4295-5G
TCIAD4295-1GEA 1125 NOK
TCIAD4295-1G TCIAD4295-5G
5,6-Dichlorobenzimidazole Hydrochloride ≥98.0% (by HPLC, titration analysis)
5,6-Dichlorobenzimidazole Hydrochloride
Formel: C₇H₄Cl₂N₂
MW: 223,48 g/mol
MDL nummer: MFCD16810292
CAS-nummer: 1087737-96-9
Bestill nå
Spesifikasjon Testresultater
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %