Di-tert-butyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Di-tert-butyl Ester

D4311-1G D4311-5G
TCIAD4311-1GEA 690 NOK
TCIAD4311-1G TCIAD4311-5G
Di-tert-butyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, titration analysis)
Di-tert-butyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
Formel: C₁₇H₂₇NO₄
MW: 309,41 g/mol
MDL nummer: MFCD01882057
CAS-nummer: 55536-71-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR