2,3-Dibromo-5-chloropyridine ≥98.0% (by GC)

Leverandør: TCI
Warning

D4381-5G D4381-25G
TCIAD4381-5GEA 650 NOK
TCIAD4381-5G TCIAD4381-25G
2,3-Dibromo-5-chloropyridine ≥98.0% (by GC)
2,3-Dibromo-5-chloropyridine
Formel: C₅H₂Br₂ClN
MW: 271,3400 g/mol
MDL nummer: MFCD08687254
CAS-nummer: 137628-17-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR