6,8-Dichloro-3-formylchromone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
6,8-Dichloro-3-formylchromone, 6,8-Dichlorochromone-3-carboxaldehyde, 6,8-Dichloro-4-oxo-4H-chromene-3-carbaldehyde, 6,8-Dichloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde
D4388-1G
D4388-5G
TCIAD4388-1GEA
715
NOK
TCIAD4388-1G
TCIAD4388-5G
6,8-Dichloro-3-formylchromone ≥98.0% (by GC)
6,8-Dichloro-3-formylchromone
Formel:
C₁₀H₄Cl₂O₃ MW: 243,0430 g/mol |
MDL nummer:
MFCD00051510 CAS-nummer: 64481-10-3 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 170.0 to 174.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...