Dicyclopentyl(dimethoxy)silane ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Dicyclopentyldimethoxysilane

D4396-25G D4396-5G
TCIAD4396-25GEA 1525 NOK
TCIAD4396-25G TCIAD4396-5G
Dicyclopentyl(dimethoxy)silane ≥98.0% (by GC)
Dicyclopentyl(dimethoxy)silane
Formel: C₁₂H₂₄O₂Si
MW: 228,41 g/mol
Kokepunkt: 120°C
MDL nummer: MFCD01074471
CAS-nummer: 126990-35-0
EINECS: 404-370-8
FN: 3082
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR