2,2-Dichlorocyclopropylbenzene ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 1,1-Dichloro-2-phenylcyclopropane, (2,2-Dichlorocyclopropyl)benzene

D4410-5G D4410-25G
TCIAD4410-5GEA 260 NOK
TCIAD4410-5G TCIAD4410-25G
2,2-Dichlorocyclopropylbenzene ≥98.0% (by GC)
2,2-Dichlorocyclopropylbenzene
Formel: C₉H₈Cl₂
MW: 187,07 g/mol
Kokepunkt: 105°C
Flash punkt: 104°C
MDL nummer: MFCD00001272
CAS-nummer: 2415-80-7
EINECS: 219-326-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR