9,10-Dihydro-9,10-[3,4]thiophenoanthracene ≥98.0% (by GC)

Leverandør: TCI

D4412-200MG D4412-1G
TCIAD4412-200MGEA 1750 NOK
TCIAD4412-200MG TCIAD4412-1G
9,10-Dihydro-9,10-[3,4]thiophenoanthracene ≥98.0% (by GC)
9,10-Dihydro-9,10-[3,4]thiophenoanthracene
Formel: C₁₈H₁₂S
Smeltepunkt: 268°C
CAS-nummer: 42490-26-6

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR