N,N'-Dihydroxypyromellitimide ≥96.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: NDHPI, N,N'-Dihydroxypyromellitic Diimide, 2,6-Dihydroxypyrrolo[3,4-f]isoindole-1,3,5,7(2H,6H)-tetraone

D4413-1G D4413-5G
TCIAD4413-1GEA 1400 NOK
TCIAD4413-1G TCIAD4413-5G
N,N'-Dihydroxypyromellitimide ≥96.0% (by HPLC, total nitrogen)
N,N'-Dihydroxypyromellitimide
Formel: C₁₀H₄N₂O₆
MW: 248.15 g/mol
MDL nummer: MFCD28334210
CAS-nummer: 57583-53-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR