N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide ≥92.0% (by total nitrogen basis)

Leverandør: TCI
Warning

Synonymer: Pigment Red 179

D4429-1G D4429-5G
TCIAD4429-1GEA 1225 NOK
TCIAD4429-1G TCIAD4429-5G
N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide ≥92.0% (by total nitrogen basis)
N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide
Formel: C₂₆H₁₄N₂O₄
MW: 418.41 g/mol
MDL nummer: MFCD00071975
CAS-nummer: 5521-31-3
EINECS: 226-866-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a001279b27608eef76a4a63fcd3c91e013597b6d&vid=a001279b27608eef76a4a63fcd3c91e013597b6d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Purity (with Total Nitrogen) min. 92.0 %
Lambda max. 590.0 to 600.0 nm (H2SO4)
Molar extinction coefficient min.76000 (H2SO4, 590.0 to 600.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR