4-Decenoic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-Decenoic Acid, 4-Decenoic acid

D4449-5ML D4449-25ML
TCIAD4449-5MLEA 970 NOK
TCIAD4449-5ML TCIAD4449-25ML
4-Decenoic acid ≥98.0% (by GC, titration analysis)
4-Decenoic acid
Formel: C₁₀H₁₈O₂
MW: 170,25 g/mol
MDL nummer: MFCD03092963
CAS-nummer: 26303-90-2
EINECS: 247-599-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR