3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Warning

Synonymer: DOC Iodide, 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzoxazolylidene]-1-propenyl]benzoxazolium Iodide, 3,3'-Diethyloxacarbocyanine iodide, DiOC2(3) iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]oxazol-3-ium iodide

D4453-1G D4453-5G
TCIAD4453-1GEA 1100 NOK
TCIAD4453-1G TCIAD4453-5G
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥98.0% (by total nitrogen basis)
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide]
Formel: C₂₁H₂₁IN₂O₂
MW: 460,32 g/mol
Smeltepunkt: 276°C
MDL nummer: MFCD00011952
CAS-nummer: 905-96-4
EINECS: 212-996-6

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark red powder to crystaline
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR