4-(2,2-Diphenylethenyl)-N,N-di(p-tolyl)aniline ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

D4478-5G D4478-25G
TCIAD4478-5GEA 1150 NOK
TCIAD4478-5G TCIAD4478-25G
4-(2,2-Diphenylethenyl)-N,N-di(p-tolyl)aniline ≥98.0% (by HPLC, total nitrogen)
4-(2,2-Diphenylethenyl)-N,N-di(p-tolyl)aniline
Formel: C₃₄H₂₉N
MW: 451,61 g/mol
MDL nummer: MFCD09832972
CAS-nummer: 89114-91-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR