3,6-Dibromo-9-n-octylcarbazole ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3,6-Dibromo-9-octyl-9H-carbazole
D4482-200MG
D4482-1G
TCIAD4482-200MGEA
580
NOK
TCIAD4482-200MG
TCIAD4482-1G
3,6-Dibromo-9-n-octylcarbazole ≥98.0% (by GC)
3,6-Dibromo-9-n-octylcarbazole
Formel:
C₂₀H₂₃Br₂N MW: 437,22 g/mol |
MDL nummer:
MFCD23703112 CAS-nummer: 79554-93-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 81.0 to 85.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...