3,4-Dimethylbenzylamine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
D4497-5ML
D4497-25ML
TCIAD4497-5MLEA
1000
NOK
TCIAD4497-5ML
TCIAD4497-25ML
3,4-Dimethylbenzylamine ≥98.0% (by GC, titration analysis)
3,4-Dimethylbenzylamine
Formel:
C₉H₁₃N MW: 135,21 g/mol Kokepunkt: 50°C |
MDL nummer:
MFCD00025577 CAS-nummer: 102-48-7 EINECS: 203-034-6 FN: 2735 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...