2,5-Dibromo-3-decylthiophene ≥97.0% (by GC)
Leverandør: TCI
D4572-25G
D4572-5G
TCIAD4572-25GEA
4500
NOK
TCIAD4572-25G
TCIAD4572-5G
2,5-Dibromo-3-decylthiophene ≥97.0% (by GC)
2,5-Dibromo-3-decylthiophene
Formel:
C₁₄H₂₂Br₂S MW: 382,20 g/mol Kokepunkt: 148°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00274314 CAS-nummer: 158956-23-1 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...